siRNA Details
| Gene Symbol: Ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177602.1 | NP_001171073.1 | 6 | 1833 | ||
| NM_001177604.1 | NP_001171075.1 | 6 | 1646 | |||
| NM_001177605.1 | NP_001171076.1 | 6 | 1624 | |||
| NM_009647.5 | NP_033777.1 | 5 | 1591 | |||
| GenBank mRNA | AK134600.1 | - | 6 | 1836 | ||
| AK144439.1 | - | 5 | 1561 | |||
| AK154374.1 | - | 6 | 1650 | |||
| AK166975.1 | - | 5 | 1514 | |||
| BC086663.1 | - | 5 | 1539 | 30649610 | ||
| BC086791.1 | - | 5 | 1554 | |||
| D85036.1 | - | 5 | 1561 | |||

Ak4
Genomic Map