siRNA Details
| Gene Symbol: RYK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005861.2 | NP_001005861.1 | 2 | 358 | ||
| NM_002958.3 | NP_002949.2 | 2 | 358 | |||
| GenBank mRNA | CN309980.1 | - | 2 | 202 | ||
| S59184.1 | - | 2 | 391 | |||
| X69970.1 | - | 2 | 358 | |||
| X96588.1 | - | 3 | 263 | |||

RYK
Genomic Map