siRNA Details
| Gene Symbol: Vmn2r52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105191.1 | NP_001098661.1 | 2 | 440 | ||
| Gene Symbol: Vmn2r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081405.1 | NP_001074874.1 | 2 | 440 | ||
More Information
Set Membership: |
|

Vmn2r52
Genomic Map