siRNA Details
| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172806.2 | NP_766394.2 | 12 | 3266 | ||
| GenBank mRNA | AK039845.1 | - | 2 | 297 | ||
| BC041669.1 | - | 12 | 3266 | 3482657 | ||
| BC057303.1 | - | 11 | 2985 | 5702276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd7| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172806.2 | NP_766394.2 | 12 | 3266 | ||
| GenBank mRNA | AK039845.1 | - | 2 | 297 | ||
| BC041669.1 | - | 12 | 3266 | 3482657 | ||
| BC057303.1 | - | 11 | 2985 | 5702276 | ||