siRNA Details
| Gene Symbol: Lamtor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019920.2 | NP_064304.1 | 5 | 357 | ||
| GenBank mRNA | AF082526.1 | - | 5 | 357 | ||
| AK031303.1 | - | 1 | 743 | |||
| AK088454.1 | - | 5 | 364 | |||
| AK151432.1 | - | 5 | 387 | |||
| AK159450.1 | - | 5 | 355 | |||
| AK162102.1 | - | 5 | 349 | |||
| BC016417.1 | - | 5 | 340 | 3158151 | ||
| BC083155.1 | - | 5 | 339 | 5339040 | ||

Lamtor3
Genomic Map