siRNA Details
| Gene Symbol: Vmn2r110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104572.1 | NP_001098042.1 | 3 | 876 | ||
| Gene Symbol: Vmn2r108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104570.1 | NP_001098040.1 | 3 | 876 | ||
More Information
Set Membership: |
|

Vmn2r110
Genomic Map