siRNA Details
| Gene Symbol: Slc5a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145423.2 | NP_663398.2 | 5 | 918 | ||
| GenBank mRNA | AK039927.1 | - | 5 | 747 | ||
| AK136457.1 | - | 5 | 928 | |||
| AY484428.1 | - | 5 | 918 | |||
| BC017691.1 | - | 873 | 4239161 | |||

Slc5a8
Genomic Map