siRNA Details
| Gene Symbol: 4932414J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030001.1 | - | 2 | 1216 | ||
| BC109353.1 | - | 1 | 1007 | 40057015 | ||
| BC116680.1 | - | 1 | 1007 | 40106469 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932414J04Rik| Gene Symbol: 4932414J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030001.1 | - | 2 | 1216 | ||
| BC109353.1 | - | 1 | 1007 | 40057015 | ||
| BC116680.1 | - | 1 | 1007 | 40106469 | ||