siRNA Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173364.5 | NP_775540.4 | 2 | 478 | ||
| GenBank mRNA | AY341877.1 | - | 1 | 319 | ||
| BC099896.1 | - | 1 | 290 | 30545532 | ||
| BC099930.1 | - | 2 | 423 | 30058096 | ||
| CD559799.1 | - | 2 | 413 | |||

Zfp445
Genomic Map