siRNA Details
Gene Symbol: 4930503E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029131.3 | NP_083407.1 | 2 | 300 | ||
GenBank mRNA | AK015686.1 | - | 2 | 300 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930503E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029131.3 | NP_083407.1 | 2 | 300 | ||
GenBank mRNA | AK015686.1 | - | 2 | 300 |