siRNA Details
| Gene Symbol: Efcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159697.1 | NP_001153169.1 | 8 | 2361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcc1| Gene Symbol: Efcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159697.1 | NP_001153169.1 | 8 | 2361 | ||