siRNA Details
| Gene Symbol: S100A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024210.1 | NP_001019381.1 | 5 | 697 | ||
| NM_001024211.1 | NP_001019382.1 | 3 | 413 | |||
| NM_001024212.1 | NP_001019383.1 | 3 | 384 | |||
| NM_001024213.1 | NP_001019384.1 | 3 | 352 | |||
| NM_005979.2 | NP_005970.1 | 4 | 489 | |||
| XM_005245434.3 | XP_005245491.1 | 5 | 604 | |||
| XM_011509862.2 | XP_011508164.1 | 7 | 904 | |||
| XM_011509863.2 | XP_011508165.1 | 7 | 871 | |||
| XM_011509864.1 | XP_011508166.1 | 5 | 694 | |||
| XM_017002034.1 | XP_016857523.1 | 6 | 787 | |||
| XM_017002035.1 | XP_016857524.1 | 6 | 788 | |||
| XM_017002036.1 | XP_016857525.1 | 3 | 521 | |||
| GenBank mRNA | AK097132.1 | - | 5 | 1993 | ||
| AY987392.1 | - | 2 | 190 | |||
| BC000632.2 | - | 2 | 262 | 3343203 | ||
| BC068064.1 | - | 2 | 248 | 6074799 | ||
| BC070291.1 | - | 4 | 489 | 6713909 | ||
| BM762088.1 | - | 3 | 386 | |||
| BM913375.1 | - | 556 | ||||
| BT006724.1 | - | 2 | 193 | |||
| CR542149.1 | - | 2 | 193 | |||
| X99920.1 | - | 3 | 266 | |||

S100A13
Genomic Map