siRNA Details
| Gene Symbol: Fntb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145927.2 | NP_666039.1 | 2, 3 | 254 | ||
| GenBank mRNA | AK089911.1 | - | 2, 3 | 284 | ||
| BC031417.1 | - | 2, 3 | 254 | 3154478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fntb| Gene Symbol: Fntb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145927.2 | NP_666039.1 | 2, 3 | 254 | ||
| GenBank mRNA | AK089911.1 | - | 2, 3 | 284 | ||
| BC031417.1 | - | 2, 3 | 254 | 3154478 | ||