siRNA Details
| Gene Symbol: Gm5891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034904.2 | NP_001030076.1 | 4 | 756 | ||
| GenBank mRNA | AK139650.1 | - | 4 | 756 | ||
| AK163219.1 | - | 627 | ||||
| Gene Symbol: Gm10662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201364.1 | NP_001188293.1 | 4 | 754 | ||

Gm5891
Genomic Map