siRNA Details
| Gene Symbol: SALL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291446.1 | NP_001278375.1 | 3 | 1409 | ||
| NM_001291447.1 | NP_001278376.1 | 3 | 1389 | |||
| NM_005407.2 | NP_005398.2 | 2 | 1814 | |||
| XM_011537064.1 | XP_011535366.1 | 2 | 1740 | |||
| XM_011537065.1 | XP_011535367.1 | 2 | 1886 | |||
| GenBank mRNA | AB002358.2 | - | 2 | 1801 | ||
| AK293574.1 | - | 3 | 1095 | |||
| AK296414.1 | - | 3 | 1208 | |||
| AK299931.1 | - | 2 | 1688 | |||
| BC136528.1 | - | 2 | 1549 | 9020518 | ||
| BC136529.1 | - | 2 | 1549 | 9020519 | ||
| X98834.1 | - | 2 | 1701 | |||

SALL2
Genomic Map