siRNA Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005981.3 | NP_005972.1 | 3 | 453 | ||
| XM_005269074.3 | XP_005269131.2 | 3 | 1378 | |||
| GenBank mRNA | AK090502.1 | - | 2 | 525 | ||
| AK294128.1 | - | 3 | 406 | |||
| AK315765.1 | - | 3 | 574 | |||
| BC010377.1 | - | 3 | 406 | 4294627 | ||
| CR407647.1 | - | 3 | 299 | |||
| U01160.1 | - | 3 | 401 | |||

TSPAN31
Genomic Map