siRNA Details
| Gene Symbol: Nrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013372.2 | NP_001013390.1 | 1 | 156 | ||
| GenBank mRNA | AY753183.1 | - | 1 | 156 | ||
| BC119399.1 | - | 1 | 133 | 8734148 | ||
| BC119401.1 | - | 1 | 133 | 8734150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrp| Gene Symbol: Nrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013372.2 | NP_001013390.1 | 1 | 156 | ||
| GenBank mRNA | AY753183.1 | - | 1 | 156 | ||
| BC119399.1 | - | 1 | 133 | 8734148 | ||
| BC119401.1 | - | 1 | 133 | 8734150 | ||