siRNA Details
| Gene Symbol: Defb33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039119.2 | NP_001034208.1 | 3 | 264 | ||
| GenBank mRNA | BC145764.1 | - | 2 | 202 | 40131067 | |
| BC145766.1 | - | 2 | 202 | 40131069 | ||
| BY092288.1 | - | 3 | 265 | |||
| BY461224.1 | - | 3 | 283 | |||
| DQ012041.1 | - | 2 | 113 | |||

Defb33
Genomic Map