siRNA Details
| Gene Symbol: Kncn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039124.3 | NP_001034213.1 | 3 | 570 | ||
| GenBank mRNA | BC132034.1 | - | 3 | 517 | 40130311 | |
| BC132036.1 | - | 3 | 517 | 40130313 | ||
| BC145375.1 | - | 3 | 547 | 9053933 | ||
| DQ235452.1 | - | 3 | 570 | |||

Kncn
Genomic Map