siRNA Details
| Gene Symbol: Defb47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039125.2 | NP_001034214.1 | 2 | 125 | ||
| GenBank mRNA | BC132604.1 | - | 2 | 125 | 40130881 | |
| BC132606.1 | - | 2 | 125 | 40130883 | ||
| DQ012043.1 | - | 2 | 100 | |||

Defb47
Genomic Map