siRNA Details
| Gene Symbol: Zfp784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039532.2 | NP_001034621.1 | 2 | 194 | ||
| GenBank mRNA | AK050452.1 | - | 2 | 189 | ||
| AK161791.1 | - | 2 | 196 | |||
| BB620617.1 | - | 2 | 195 | |||
| BC147237.1 | - | 2 | 181 | 9056299 | ||
| BC147238.1 | - | 2 | 181 | 9056303 | ||

Zfp784
Genomic Map