siRNA Details
| Gene Symbol: Angptl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039554.3 | NP_001034643.1 | 4 | 850 | ||
| GenBank mRNA | AK031675.1 | - | 4 | 850 | ||
| BC023373.1 | - | 4 | 837 | 5043159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Angptl7| Gene Symbol: Angptl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039554.3 | NP_001034643.1 | 4 | 850 | ||
| GenBank mRNA | AK031675.1 | - | 4 | 850 | ||
| BC023373.1 | - | 4 | 837 | 5043159 | ||