siRNA Details
| Gene Symbol: Smco3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039558.2 | NP_001034647.2 | 2 | 406 | ||
| GenBank mRNA | AK045581.1 | - | 2 | 383 | ||
| AK047766.1 | - | 2 | 575 | |||
| BB624054.1 | - | 2 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smco3| Gene Symbol: Smco3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039558.2 | NP_001034647.2 | 2 | 406 | ||
| GenBank mRNA | AK045581.1 | - | 2 | 383 | ||
| AK047766.1 | - | 2 | 575 | |||
| BB624054.1 | - | 2 | 406 | |||