siRNA Details
| Gene Symbol: Cypt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039941.2 | NP_001035030.1 | 1 | 285 | ||
| GenBank mRNA | AM040457.1 | - | 1 | 285 | ||
| Gene Symbol: Cypt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016467.1 | - | 1 | 293 | ||
| AM040451.1 | - | 1 | 285 | |||

Cypt8
Genomic Map