siRNA Details
| Gene Symbol: Cypt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039943.2 | NP_001035032.1 | 1, 2 | 289 | ||
| GenBank mRNA | AM040456.1 | - | 1, 2 | 289 | ||
| Gene Symbol: Cypt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF463502.1 | - | 1, 2 | 276 | ||
| AK006892.1 | - | 1, 2 | 298 | |||
| AM040454.1 | - | 1, 2 | 289 | |||
| AM040455.1 | - | 1, 2 | 289 | |||
| BC048455.1 | - | 1, 2 | 294 | |||
| BC061079.1 | - | 1, 2 | 294 | 6773161 | ||

Cypt7
Genomic Map