siRNA Details
| Gene Symbol: Dux | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081954.1 | NP_001075423.1 | 464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dux| Gene Symbol: Dux | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081954.1 | NP_001075423.1 | 464 | |||