siRNA Details
| Gene Symbol: Nova1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021361.1 | NP_067336.1 | 2 | 238 | ||
| GenBank mRNA | AF232828.1 | - | 2 | 172 | ||
| AK090295.1 | - | 2 | 527 | |||
| BM942596.1 | - | 2 | 238 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001100541, XM_006240070, XM_006240069.

Nova1
Genomic Map