siRNA Details
| Gene Symbol: Clec2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101507.1 | NP_001094977.1 | 3 | 515 | ||
| GenBank mRNA | CJ059176.1 | - | 3 | 306 | ||
| CJ059317.1 | - | 2 | 113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec2l| Gene Symbol: Clec2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101507.1 | NP_001094977.1 | 3 | 515 | ||
| GenBank mRNA | CJ059176.1 | - | 3 | 306 | ||
| CJ059317.1 | - | 2 | 113 | |||