siRNA Details
| Gene Symbol: Gm7628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083067.1 | - | 1 | 1197 | ||
| AK142995.1 | - | 2 | 1267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7628| Gene Symbol: Gm7628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083067.1 | - | 1 | 1197 | ||
| AK142995.1 | - | 2 | 1267 | |||