siRNA Details
| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008997.3 | NP_033023.1 | 3 | 491 | ||
| GenBank mRNA | AB232606.1 | - | 3 | 267 | ||
| AK134125.1 | - | 3 | 236 | |||
| AK146452.1 | - | 3 | 352 | |||
| AK146460.1 | - | 3 | 303 | |||
| AK146623.1 | - | 3 | 303 | |||
| AK146978.1 | - | 3 | 283 | |||
| AK147077.1 | - | 3 | 322 | |||
| AK148022.1 | - | 3 | 364 | |||
| AK148106.1 | - | 3 | 290 | |||
| AK177220.1 | - | 3 | 208 | |||
| AK188436.1 | - | 3 | 258 | |||
| AK195913.1 | - | 2 | 147 | |||
| BC054753.1 | - | 3 | 307 | 6411651 | ||
| BC085270.1 | - | 3 | 300 | 6478784 | ||
| BC157916.1 | - | 3 | 320 | 9007344 | ||
| BU705914.1 | - | 3 | 491 | |||
| KJ650201.1 | - | 3 | 267 | |||
| L26528.1 | - | 3 | 302 | |||

Rab11b
Genomic Map