siRNA Details
Gene Symbol: Gm7714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110779.1 | NP_001104249.1 | 3 | 191 | ||
GenBank mRNA | AK137735.1 | - | 3 | 193 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110779.1 | NP_001104249.1 | 3 | 191 | ||
GenBank mRNA | AK137735.1 | - | 3 | 193 |