siRNA Details
| Gene Symbol: Gm16513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200055.1 | NP_001186984.1 | 2542 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16513| Gene Symbol: Gm16513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200055.1 | NP_001186984.1 | 2542 | |||