siRNA Details
| Gene Symbol: 4921528I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014978.1 | - | 10, 11 | 723 | ||
| AK077029.1 | - | 10, 11 | 724 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921528I07Rik| Gene Symbol: 4921528I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014978.1 | - | 10, 11 | 723 | ||
| AK077029.1 | - | 10, 11 | 724 | |||