siRNA Details
| Gene Symbol: 4921528I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014978.1 | - | 12 | 1012 | ||
| AK077029.1 | - | 12 | 1014 | |||
| Gene Symbol: 4930579C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029815.1 | - | 12 | 1009 | ||

4921528I07Rik
Genomic Map