siRNA Details
Gene Symbol: A430089I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177913.4 | NP_808581.1 | 4 | 1788 |
Gene Symbol: BC080696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC080696.1 | - | 3 | 1713 |
