siRNA Details
| Gene Symbol: Gsdmcl-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016931.1 | - | 876 | |||
| Gene Symbol: Gsdmcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019629.1 | - | 832 | |||

Gsdmcl-ps
Genomic Map