siRNA Details
| Gene Symbol: Krtap4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048196.1 | NP_001041661.1 | 1 | 856 | ||
| GenBank mRNA | AK137752.1 | - | 1 | 857 | ||
| BC147087.1 | - | 1 | 827 | 9056137 | ||
| BC147088.1 | - | 1 | 827 | 9056139 | ||

Krtap4-1
Genomic Map