siRNA Details
Gene Symbol: Krtap4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048196.1 | NP_001041661.1 | 1 | 967 | ||
GenBank mRNA | AK137752.1 | - | 1 | 968 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048196.1 | NP_001041661.1 | 1 | 967 | ||
GenBank mRNA | AK137752.1 | - | 1 | 968 |