siRNA Details
Gene Symbol: Vmn2r114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102584.1 | NP_001096054.1 | 4 | 1331 | ||
GenBank mRNA | AB540945.1 | - | 4 | 1331 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
