siRNA Details
| Gene Symbol: Frmpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081172.2 | NP_001074641.1 | 16 | 3754 | ||
| GenBank mRNA | AK122409.1 | - | 13 | 3383 | ||
| AK133351.1 | - | 3 | 2193 | |||
| BC031840.1 | - | 6 | 2614 | |||
| BC046469.1 | - | 2 | 1530 | |||
| BC051097.1 | - | 6 | 2614 | |||
| BC158060.1 | - | 16 | 3718 | 9007360 | ||

Frmpd1
Genomic Map