siRNA Details
| Gene Symbol: A430089I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177913.4 | NP_808581.1 | 4 | 1668 | ||
| GenBank mRNA | AK040373.1 | - | 4 | 1669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A430089I19Rik| Gene Symbol: A430089I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177913.4 | NP_808581.1 | 4 | 1668 | ||
| GenBank mRNA | AK040373.1 | - | 4 | 1669 | ||