siRNA Details
| Gene Symbol: Gm8013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA166375.1 | - | 2 | 172 | ||
| AI509282.1 | - | 2 | 172 | |||
| BX527816.1 | - | 1 | 51 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8013| Gene Symbol: Gm8013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA166375.1 | - | 2 | 172 | ||
| AI509282.1 | - | 2 | 172 | |||
| BX527816.1 | - | 1 | 51 | |||