siRNA Details
| Gene Symbol: Dspp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010080.2 | NP_034210.2 | 4 | 506 | ||
| GenBank mRNA | BC116173.1 | - | 507 | |||
| BC129802.1 | - | 506 | 40049023 | |||
| U67916.1 | - | 4 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dspp| Gene Symbol: Dspp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010080.2 | NP_034210.2 | 4 | 506 | ||
| GenBank mRNA | BC116173.1 | - | 507 | |||
| BC129802.1 | - | 506 | 40049023 | |||
| U67916.1 | - | 4 | 515 | |||