siRNA Details
| Gene Symbol: Zfp541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099277.1 | NP_001092747.1 | 13 | 3581 | ||
| GenBank mRNA | BC157962.1 | - | 11 | 3419 | 9007287 | |
| DQ864732.1 | - | 13 | 3581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp541| Gene Symbol: Zfp541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099277.1 | NP_001092747.1 | 13 | 3581 | ||
| GenBank mRNA | BC157962.1 | - | 11 | 3419 | 9007287 | |
| DQ864732.1 | - | 13 | 3581 | |||