siRNA Details
| Gene Symbol: Zfp781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199062.1 | NP_951017.1 | 4 | 422 | ||
| GenBank mRNA | BC062127.1 | - | 4 | 422 | 6827033 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp781| Gene Symbol: Zfp781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199062.1 | NP_951017.1 | 4 | 422 | ||
| GenBank mRNA | BC062127.1 | - | 4 | 422 | 6827033 | |