siRNA Details
Gene Symbol: Zfp781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199062.1 | NP_951017.1 | 4 | 348 | ||
GenBank mRNA | BC062127.1 | - | 4 | 348 | 6827033 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199062.1 | NP_951017.1 | 4 | 348 | ||
GenBank mRNA | BC062127.1 | - | 4 | 348 | 6827033 |