siRNA Details
| Gene Symbol: Samd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081415.1 | NP_001074884.1 | 2 | 1168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | W83690.1 | - | 87 | Chr. - - [-] on Build | ||

Samd1
Genomic Map