siRNA Details
| Gene Symbol: Gm8273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084737.1 | - | 1 | 435 | ||
| BB479211.2 | - | 1 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8273| Gene Symbol: Gm8273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084737.1 | - | 1 | 435 | ||
| BB479211.2 | - | 1 | 446 | |||