siRNA Details
| Gene Symbol: SRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098814.1 | NP_001092284.1 | 4 | 365 | ||
| NM_001323667.1 | NP_001310596.1 | 5 | 495 | |||
| NM_001323668.1 | NP_001310597.1 | 5 | 1743 | |||
| XM_017023527.1 | XP_016879016.1 | 5 | 1690 | |||
| XM_017023528.1 | XP_016879017.1 | 4 | 396 | |||
| XM_017023529.1 | XP_016879018.1 | 4 | 411 | |||
| GenBank mRNA | AK056588.1 | - | 4 | 365 | ||
| AK302601.1 | - | 4 | 401 | |||
| AL831973.2 | - | 4 | 337 | |||
| AL832361.1 | - | 5 | 1737 | |||
| BX647476.1 | - | 5 | 1737 | |||
| DB149163.1 | - | 4 | 365 | |||

SRL
Genomic Map