siRNA Details
| Gene Symbol: Gm8327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB030205.1 | - | 1 | 571 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8327| Gene Symbol: Gm8327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB030205.1 | - | 1 | 571 | ||